Descrizione
Luer fittings allow for a simple yet secure connection. Ideal for capping off fluid transfer system. Quickly connect and disconnect with slip luer design
Specifica
Specifica
| Certificazione/Conformità | Non-DEHP;Non-Phthalate;RoHS |
| Ingresso | Male Luer Slip, Plug, Closed at Grip |
| Materiale (guarnizione) | Nylon |