missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Maleate, 0.2M buffer soln., pH 5.0, Thermo Scientific™
Click to view available options
Quantità:
250 mL
500 mL
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Materiale o nome chimico | Maleate buffer soln. |
Colore | Colorless |
Concentrazione | 0.2 M |
pH | 5.0 |
Forma fisica | Liquid |
Quantità | 250 mL |
IUPAC Name | sodium;(Z)-4-hydroxy-4-oxobut-2-enoate;trihydrate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'