Learn More
Cytiva Manifold Tray for Ettan™ IPGphor™ 3 Isoelectric Focusing Unit
Product Code. 10166674 Shop All Cytiva Products
Change view
Click to view available options
For Use With (Equipment):
Ettan IPGphor 3 IEF Unit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
10166674 Ettan IPGphor 3 IEF Unit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10166674 Supplier Cytiva Supplier No. 80649857
Spedizione stimata: 08-01-2027
Manifold Tray for Ettan™ IPGphor™ 3 Isoelectric Focusing Unit
  • Uniform thermal characteristics
  • High heat conductivity
  • No wetted part
  • Sample equilibration in same tray
  • No. of gels: 1 to 12
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Ettan IPGphor 3 IEF Unit
Type IPGphor Manifold Ceramic Tray
Material Ceramic
No. of Gels 1 to 12
Quantity Complete packsize