Learn More
Thermo Scientific™ Mannitol Salt Agar (Dehydrated)
Codice prodotto. 10269532
Cambia vista
Click to view available options
Quantità:
Confezioni singole
2.5kg
5 kg
Dimensione della confezione:
3kg
500g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
10269532 5 kg 5kg
10096012 Confezioni singole 500g
12774136 2.5kg 3kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Codice prodotto. 10269532 Fornitore Thermo Scientific™ N. del fornitore CM0085T
Please to purchase this item. Need a web account? Register with us today!
Presumptively detect, isolate and enumerate pathogenic staphylococci in milk, food, fish and other specimens with this highly selective medium.
Presumptively detect, isolate and enumerate pathogenic staphylococci in milk, food, fish and other specimens with Thermo Scientific™ Oxoid™ Mannitol Salt Agar (Dehydrated). Due to high concentrations of salt in the medium, most unwanted bacteria are inhibited with the exception of some halophilic marine organisms. Presumptive coagulase-positive staphylococci produce colonies surrounded by bright yellow zones while non-pathogenic staphylococci produce colonies with reddish purple zones.
Staphylococcus aureus is a coagulase-positive, catalase-positive, Gram-positive cocci that occur singly or in pairs, tetrads, short chains, and grape-like clusters. S. aureus can produce a variety of syndromes with clinical manifestations, including skin and soft tissue infections, empyema, bloodstream infections, pneumonia, osteomyelitis, septic arthritis, endocarditis, sepsis, and meningitis.
Clearly and easily differentiate Staphylococcus aureus and Staphylococcus epidermidis while most other bacteria is inhibited by the high salt concentration.
  • Highly selective: due to high concentrations of salt in the medium.
  • Easy to differentiate: Staphylococcus aureus forms yellow colonies with yellow halo while Staphylococcus epidermis forms pink colonies with medium turning pink.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand
TRUSTED_SUSTAINABILITY
Specifica
Forma Powder
Product Type Mannitol Salt Agar
Quantità 5 kg
For Laboratory Use Only