Learn More
Welch™ Manual Ball Valve
Product Code. 10576122 Shop All Welch Ilmvac Products
Change view
Click to view available options
Type:
Ball Valve
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
10576122 Ball Valve Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10576122 Supplier Welch™ Supplier No. 700048
  • ONLINE EXCLUSIVE PRICE
    224.00€
    212.08€ / pezzi 1
    Risparmia 11.92€
    5% Sconto
Spedizione stimata: 11-12-2026
Use these two-way valves in a wide variety of high pressure applications. Welch Ilmvac™ Manual Ball Valves provide full unrestricted flow that allows no product restrictions through the valve.
  • Two-way ball valve, DN 25
  • Brass nickel-plated
  • Manually operated
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) High pressure applications
Quantity 1 Each
Type Ball Valve