Learn More
Thermo Scientific™ Manual Crimpers and Decrimpers for Sealing and Unsealing Crimp Vials
Product Code. 17354163 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
11 mm Crimp Vials
13 mm Crimp Vials
20 mm Crimp Vials
8 mm Crimp Vials
Type:
Manual Crimper
Manual Decrimper
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Type Unit Size
17354163 8 mm Crimp Vials Manual Crimper Each
17314163 11 mm Crimp Vials Manual Crimper Each
17324163 13 mm Crimp Vials Manual Crimper Each
17334163 20 mm Crimp Vials Manual Crimper Each
17364163 11 mm Crimp Vials Manual Decrimper Each
17374163 13 mm Crimp Vials Manual Decrimper Each
17383763 20 mm Crimp Vials Manual Decrimper Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 17354163 Supplier Thermo Scientific™ Supplier No. 60180CR8
Spedizione stimata: 02-12-2026
Use Thermo Scientific™ Manual Vial Crimpers and Decrimpers to attach or remove crimp caps from vials, respectively.
Use Thermo Scientific™ Vial Crimpers and Decrimpers to manually attach or remove crimp caps from vials, respectively. These manual crimpers and decappers are available in 8 mm, 11 mm, 13 mm, and 20 mm sizes and allow you to easily seal or remove crimp caps reliably.
TRUSTED_SUSTAINABILITY
Specifications
Type Manual Crimper
For Use With (Equipment) 8 mm Crimp Vials
Material Stainless Steel/Aluminum
Product Line SureSTART Level 2