Learn More
Thermo Scientific™ Manual Crimpers and Decrimpers for Sealing and Unsealing Crimp Vials
Sfoglia Tutto Thermo Scientific Prodotti
Descrizione
Use Thermo Scientific™ Vial Crimpers and Decrimpers to manually attach or remove crimp caps from vials, respectively. These manual crimpers and decappers are available in 8 mm, 11 mm, 13 mm, and 20 mm sizes and allow you to easily seal or remove crimp caps reliably.
Specifica
Specifica
| Tipo | Manual Decrimper |
| Da utilizzare con (apparecchiatura) | 11 mm Crimp Vials |
| Materiale | Stainless Steel/Aluminum |
| Linea di prodotti | SureSTART Level 2 |
| Green Features | ACT Ecolabel (Gold) |