Learn More
Innopsys MAPIX Analyzer
Intuitive and easy-to handle software for the quantification of microarray data from PEPperCHIP™ Peptide Microarrays
Marca: Innopsys MPX.001.000
Dettagli aggiuntivi : Peso : 0.30000kg
Descrizione
- For microarray image analysis and data quantification
- Intuitive and straightforward handling using .GAL files
- Available for Windows 32 and 64 bit operating systems
Specifica
Software | |
Microarray Image Analysis and Data Quantification | |
Windows Operating Systems (32/64 Bit) | |
Full Single User License |