Learn More
Dr. Ehrenstorfer Mass-Labelled PFAS-24ES Extraction Mixture 1 μg/mL in Methanol:Water
Descrizione
- Certified reference materials
Specifica
Specifica
| Materiale o nome chimico | Perfluoroundecanoic Acid 13C7 (1,2,3,4,5,6,7-13C7), 2-(N-Methylperfluorooctanesulfonamido)acetic acid D3 (N-methyl D3), 1H,1H,2H,2H-Perfluorohexanesulfonic Acid 13C2 (1,2-13C2) Sodium, 2-(N-Ethylperfluorooctanesulfonamido)acetic Acid D5 (N-Ethyl D5), Perfluorohexanoic Acid 13C5 (1,2,3,4,6-13C5), Perfluorooctanoic acid 13C8, Perfluorobutanoic acid 13C4, Perfluoroheptanoic Acid 13C4 (1,2,3,4-13C4), Perfluorononanoic acid 13C9, Perfluorotetradecanoic Acid 13C2 (1,2-13C2), Perfluorooctanesulfonic acid 13C8 sodium, Perfluorooctane Sulfonamide 13C8, Perfluoropentanoic acid 13C5, Perfluorodecanoic acid 13C6 (1,2,3,4,5,6-13C6), Perfluorobutanesulfonic Acid 13C3 (2,3,4-13C3) Sodium, Perfluorohexanesulfonic Acid 13C3 (1,2,3-13C3) Sodium, 1H,1H,2H,2H-Perfluorooctanesulfonic Acid 13C2 (1,2-13C2) Sodium, 1H,1H,2H,2H-Perfluorodecanesulfonic Acid 13C2 (1,2-13C2) Sodium, Perfluorododecanoic Acid (1,2-13C2) |
| CAS | 1400690-70-1, 2708218-88-4, 1265205-97-7, 2328024-54-8, 1350614-84-4, 1017281-29-6 |
| Solvente o matrice | Methanol/Water |
| Quantità | 1 mL |