Descrizione
Thermo Scientific MassRuler DNA Ladders are designed for accurate quantification and sizing of DNA fragments by agarose gel electrophoresis. Individuals DNA fragments are chromatography-purified and calibrated against a standard that guarantees precise quantity of each band.
Highlights
- Accurate DNA sizing and quantification – DNA ladder fragments are quantified spectrophotometrically
- Sharp bands, easy-to-remember fragment sizes and quantities
- Ready-to-use—premixed with 6X MassRuler DNA Loading Dye for direct loading
- Stable at room temperature for up to 24 months
Sustainability features
Ready-to-use MassRuler DNA ladders can be stored at room temperature for up to 24 months. The ladder is shipped at ambient temperature, using optimized right-sized packaging and 100% recyclable packaging reducing waste.
Specifica
Specifica
| Concentrazione | 6X |
| Gel Type | Agarose |
| Label or Dye | Bromophenol Blue |
| Ready to Load | Yes |
| Intervallo dimensioni | 80 to 10000 bp |
| Compatibilità gel | Agarose Gels |
| Quantità | 2 x 500 μL |
| Product Line | MassRuler |
| Product Type | DNA Ladder Mix |