Learn More
Masterflex™ I/P™ Precision Modular Pump
Codice prodotto. 13487099 Sfoglia Tutto Masterflex Prodotti
Cambia vista
Click to view available options
Include:
I/P Modular Dispensing Drive
I/P Modular Washdown Drive
Dimensione della confezione:
Pezzo
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Include Dimensione della confezione
13487099 I/P Modular Washdown Drive Pezzo
13477099 I/P Modular Dispensing Drive Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Codice prodotto. 13487099 Fornitore Masterflex™ N. del fornitore 7796230
  • ONLINE EXCLUSIVE PRICE
    8285.00€
    6945.16€ / pezzi 1
    Risparmia 1339.84€
    16% Sconto
Spedizione stimata: 02-02-2027
Purge tubing before or after pumping and pump fluid in either direction. Masterflex™ I/P™ Precision Modular Pumps allow you the flexibility to separate the motor from the controller. Designed to fit in limited or awkward spaces.
  • Ideal for metering and critical transfer applications
  • These accurate modular drives have powerful motors, speed display and ±0.25% speed control for precise flow regulation
  • Splash-resistant ABS plastic controller housing is IP33 rated; drive housing is epoxy finished steel, IP33 rated
  • Powerful motors let you stack two pump heads for increased flow rates and for multichannel applications
  • Remote control capabilities via DB9 female connector on back of drive
  • Speed control input 4 to 20mA or 0 to 10V
  • Pump direction (CW/CCW) via open collector or contact closure; Start/stop via open collector or contact closure
  • Modular drive lets you place motor and controller where convenient
  • Three-digit LED display shows motor rpm
  • Membrane keypad for easy up-front control of speed, direction, and start/stop
  • Reversible motor
  • LED indicators confirm motor direction
Includes:
  • 6 ft. (1.8 m) Line Cord with IEC 320/CEE22 Socket
  • C-Flex ULTRA I/P 73 06434-73 Tubing, 10 ft. (3 m)
  • Easy-Load 77602-10 Pump Head
Specifica
Certificazione/Conformità CE
Control Type Variable Speed
Tipo di display LED
Valori nominali armadio IP66
Portata 0.6 to 8 L/min.
Motor Speed 6 to 650 rpm
Reversible Yes
Include I/P Modular Washdown Drive
Materiale Aluminum (Controller), Aluminum (Drive), Steel (Drive)
Modello I/P Easy-Load
Tipo di motore Precision washdown modular drive with wall-mount controller
Testa pompa I/P Easy-Load pump head 77602-10
Controllo della velocità ± 0.25%
Tipo di tubi C-Flex ULTRA I/P 73 Tubing, 10 ft. (3 m)
Overall Length (English) 17.25 in.
Lunghezza complessiva (sistema metrico) 361.95 mm
Overall Width (English) 7 in.
Larghezza complessiva (sistema metrico) 177.8 mm
Overall Height (English) 6.5 in.
Overall Height (Metric) 165.1 mm
Requisiti elettrici 4.4 A 115/230 VAC 50/60 Hz
Tensione 90/260 VAC
Amperaggio 4.4 A
Frequenza 50/60 Hz