Learn More
Masterflex™ Multi-Channel Large Cartridge Pump Head
Product Code. 11868443 Shop All Masterflex Products
Change view
Click to view available options
No. of Channels:
2
4
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Channels Unit Size
11868443 4 Each
11868463 2 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11868443 Supplier Masterflex™ Supplier No. 0751970
  • ONLINE EXCLUSIVE PRICE
    173.00€
    146.87€ / pezzi 1
    Risparmia 26.13€
    15% Sconto
Spedizione stimata: 22-12-2026
Use the cartridges for synchronous, multichannel pumping. MasterFlex™ L/S™ Large Cartridges feature accurate flow rates and adjustable tubing retainers to hold tubing in place.
  • Designed for use with Masterflex pump head models: 07519-05 and -06
  • Set tubing retainers to your tube size and you are ready to pump
  • On small cartridges, optimum retention is preset; simply load tubing and start pumping
  • Each cartridge accepts microbore and/or L/S precision pump tubing, please refer to table below
  • All cartridges have adjustable occlusion which maintains a bi-directional flow variation of 5%
  • Tighten occlusion for higher pressures, loosen for longer tubing life
  • Combine the input/output of two cartridges to reduce pulsation
  • Cartridges are constructed from polycarbonate and glass-filled nylon
Mix small and large cartridges on pump heads that accept both sizes; two small cartridges occupy the same space as one large cartridge
Only Silicone and C-Flex tubing formulations are recommended for size L/S 17 tubing when used in L/S cartridge pump heads.
Specifications
For Use With Microbore and Precision Tubing
No. of Channels 4
Occlusion Type Adjustable
Quantity 1 Each
Type Cartridge Pump Head