Learn More
3M™ Mat Edging Roll
Product Code. 17148650
Change view
Click to view available options
Color:
Black
Gray
Style:
High Profile
Medium Profile
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Style Unit Size
17148650 Gray Medium Profile Each
17148560 Black Medium Profile Each
17158560 Black High Profile Each
17168560 Gray High Profile Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 17148650 Supplier 3M™ Supplier No. 7000090555
  • ONLINE EXCLUSIVE PRICE
    710.00€
    427.19€ / pezzi 1
    Risparmia 282.81€
    39% Sconto
Spedizione stimata: 18-12-2026
A vinyl edging designed to match 3M™ Matting. Meets the needs of providing a smooth transition from the flooring to the matting.
  • For use on vinyl matting to provide an edge that tapers to the floor
TRUSTED_SUSTAINABILITY
Specifications
Dimensions (L x W) 30 x 0.038 m
For Use With (Application) Creating an edge that tapers to the floor
Style Medium Profile
Color Gray
Length (Metric) 30 m
Material Vinyl
Width (Metric) 0.038 m