missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Honeywell Safety Products™ Maxi Welder Cut Gloves
Click to view available options
Size:
8
9
Descrizione
- Cowhide leather/Kevlar™ construction.
- Winged thumb with yoke for flexibility and comfort.
- Seamless forefinger.
- Gauntlet type.
- Silicone-coated Kevlar™ palm for heat and cut protection.
- Kevlar™ fleece lining for heat insulation.
- Inner lining of cotton for comfort and sweat absorption.
- Water-repellent cowhide grain back.
- Excellent resistance to contact and radiant heat.
- Elastic on the back.
- Kevlar™ stitching.
- Split leather cuff to protect the forearm from mechanical hazards.
Specifica
Certificazione/Conformità | CE CATEGORY II Intermediary risks, EN 420-03 EEC Basic standards, EN 388-03 Mechanical hazards, EN 407-04 Heat hazards, EN 12477-2000 Welding hazards, EC Category PPE 2, EC Certificate Number 0075/797/162/10/12/0807, EC Attestation Number 0075/797/162/10/12/0807 |
Classificazione | Dexterity: 5 |
Area rivestita | Palm |
Materiale polso | Split Leather |
Stile di taglio | Gunn |
Coating Material | Silicone |
Cut Resistance Rating | 4 |
Materiale | Grain Leather, Dupont™ Kevlar™ Aramid Fibers |
Product Type | Welding Gloves |
Size | 9 |
Vedi altri risultati |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'