Learn More
Fisherbrand™ MaxiThermal Software
Product Code. 15348764 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Software Type:
MaxiThermal
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Software Type Unit Size
15348764 MaxiThermal Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15348764
 Restricted Item
Spedizione stimata: 08-02-2027
Add to basket
Add to basket
Requires
Standard USB 1.1 interface
MaxiThermal Software for USB Data Loggers.
  • Graphic representations are a simple X – Y plot of time versus temperature with unlimited zoom, view temperature data points on the graph, view total elapsed time, or the exact time when it was recorded
  • Graph and all data points (date, time and temperature) can be printed, as well as the ability to add a user note to the file for including information pertinent to that particular plot
  • Ability to view numeric data in a screen window and Data can be Exported as a CSV file for use in other software
  • Summary Information report showing the specific logger information, the Start and Stop Time, Duration of Recording, Measurement Interval, Highest, Lowest, Median and Average Temperatures as well as the High and Low Alarm information
  • Data from the logger is easily downloaded to your PC via a standard USB 1.1 interface
  • Compatible with Windows 98, NT, 2000, ME, XP, Vista, and Windows 7 and 8
Specifications
Type Datalogger Software
Display Type No Display
Software Type MaxiThermal
Includes MaxiThermal Software CD
For Use With (Application) Viewing and transferring recorded data from loggers to computer
For Use With (Equipment) Fisherbrand Traceable Datalogging Logger-Trac Thermometers