missing translation for 'onlineSavingsMsg'
Learn More

MDN1 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marca:  Thermo Scientific PA556225

Dettagli aggiuntivi : Peso : 0.02000kg

Codice prodotto. 15704141

  • 514.00€ / 100µL
Spedizione stimata: 11-04-2024
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

Immunogen sequence: QINEEISHLI SFCLYHTPVT PQELRDLWSL LHHQKVSPEE ITSLWSELFN SMFMSFWSST VTTNPEYWLM WNPLPGMQQR EAPKSVLDST LKG Highest antigen sequence identity to the following orthologs: Mouse - 62%, Rat - 66%.

MDN1 Polyclonal antibody specifically detects MDN1 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).
Specifica

Specifica

MDN1
Polyclonal
Unconjugated
MDN1
Dynein-related AAA-ATPase MDN1; KIAA0301; MDN1; MDN1, midasin homolog; MIDAS-containing protein; Midasin; midasin AAA ATPase 1; Rea1
Rabbit
Antigen affinity chromatography
RUO
23195
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry, Immunohistochemistry (Paraffin)
0.7 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q9NU22
MDN1
Recombinant protein corresponding to Human MDN1
100 μL
Primary
Human
Antibody
IgG
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali

Research Use Only