Learn More
Honeywell™ ME06 Shock-Absorbing Lanyard
Codice prodotto. 12674903
Cambia vista
Click to view available options
Length: 1.5 m
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto.
12674903 Length: 1.5 m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 12674903 Fornitore Honeywell™ N. del fornitore 1005279
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Highly resistant and durable 23 mm polyester webbing with a carabiner and scaffold hook.
  • Highly resistant and durable webbing
  • Includes carabiner and scaffold hook
  • Adapted for harsh working conditions and difficult environments
  • Better comfort in fall situations
  • Small-size tear-off dissipating element that reduces weight and makes lanyard easier to handle and carry.
TRUSTED_SUSTAINABILITY
Specifications
Tipi di connettore Carabiner, Scaffold Hook
Lunghezza (sistema metrico) 1.5 m
Materiale Polyester Webbing, Galvanized Steel, scaffold hook of white chromate steel