Learn More
Glassco Laboratory Equipments Measuring Cylinder Glass Class-B
Descrizione
- Low-form Class B measuring cylinders with hexagonal bases
- Available in 10 mL, 25 mL, 50 mL, 100 mL, 250 mL, 500 mL, and 1000 mL capacities
- Complies with DIN EN ISO 4788
- Made from 3.3 borosilicate glass
- Features single-metric graduations
- Printed in durable amber enamel
- Chemical resistant
Specifica
Specifica
| Materiale | Borosilicate Glass (3.3) |
| Capacità (sistema metrico) | 10 mL |
| Graduazioni | 10 mL |
| Autoclavabile | Autoclavable |
| Sterilità | Non-sterile |
| Tipo di base | Hexagonal |
| Class | Class B |
| Certificazione/Conformità | ISO 4788 |
| Product Type | Graduated Measuring Cylinder |