Learn More
Medetomidine hydrochloride, Tocris Bioscience™
Potent and highly selective α2 agonist
Marca: Tocris Bioscience 5160/10
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Potent, highly selective α2-adrenoceptor agonist (Ki values are 1.08 and 1750 nM for α2- and α1-adrenoceptors respectively). Displays greater selectivity over α1-adrenoceptors than clonidine and UK 14,304 (1620-, 220- and 300-fold respectively). Inhibits twitch response in electrically stimulated mouse vas deferens (pD2 = 9.0). Active in vivo; displays hypotensive, bradycardic, sedative, anxiolytic, hypothermic and analgesic effects. Active Isomer also available.Specifica
| Medetomidine hydrochloride | |
| C13H17ClN2 | |
| medetomidine hydrochloride, medetomidine hcl, domitor, precedex, 4-1-2,3-dimethylphenyl ethyl-1h-imidazole hydrochloride, medetomidine hydrochloride usan, +--4-alpha,2,3-trimethylbenzyl imidazole monohydrochloride, 1h-imidazole, 4-1-2,3-dimethylphenyl ethyl-, monohydrochloride | |
| CC1=C(C(=CC=C1)C(C)C2=CN=CN2)C.Cl | |
| 236.743 | |
| CHEBI:48556 |
| 86347-15-1 | |
| 10mg | |
| VPNGEIHDPSLNMU-UHFFFAOYSA-N | |
| 5-[1-(2,3-dimethylphenyl)ethyl]-1H-imidazole;hydrochloride | |
| 68601 | |
| 236.74 |