missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abcam Medroxyprogesterone 17-acetate, Progesterone Receptor Agonist, Solid, 500 mg
Description
- Molecular Weight: 386.5 Da
- Purity: >99%
- Form: Solid
- Activity: Potent progesterone receptor agonist; active in vitro and in vivo
- Biological Function: Angiostatic steroid; inhibits vascularization, collagenolysis, tumor growth, and plasmin generation
- Storage: Store at -20°C
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Tampone | 99.74% Medroxyprogesterone Acetate |
| Content And Storage | Store at -20°C. |
| Forma | Solid |
| Shipping Condition | Ambient |
| Target Molecule | Progesterone Receptor |
| Formulazione | 99.74% Medroxyprogesterone Acetate |
| Quantità | 500 mg |
| Da utilizzare con (applicazione) | Angiostatic research, tumor growth inhibition, plasmin generation inhibition |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?