Learn More
Melanocortin-1 R/MC1R Antibody (JE39-49), Novus Biologicals™
Codice prodotto. 30113455 Sfoglia Tutto Bio Techne Prodotti
Cambia vista
Click to view available options
Quantità:
100 μL
Dimensione della confezione:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
30113455 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 30113455 Fornitore Novus Biologicals N. del fornitore NBP332574
Rabbit Monoclonal Antibody
Melanocortin-1 R/MC1R Monoclonal antibody specifically detects Melanocortin-1 R/MC1R in Human,Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifica
Antigene Melanocortin-1 R/MC1R
Applicazioni Western Blot
Classificazione Monoclonal
Clone JE39-49
Concentrazione 1 mg/mL
Coniugato Unconjugated
Diluizione Western Blot 1:1000
Formulazione 1*TBS (pH7.4), 0.05% BSA and 40% Glycerol
Alias gene CMM5, MC1-R, melanocortin 1 receptor (alpha melanocyte stimulating hormone receptor), Melanocortin receptor 1, melanocyte-stimulating hormone receptor, melanotropin receptor, MSHR, MSH-RMGC14337, SHEP2
Specie ospite Rabbit
Immunogeno Synthetic peptide within Human Melanocortin-1 R/MC1R aa 1-50/ 317. (Uniprot: Q01726)
Metodo di purificazione Protein A purified
Quantità 100 μL
Status giuridico RUO
Disciplina di ricerca GPCR, Neuroscience
Primario o secondario Primary
ID gene (immissione) 4157
Target Species Human, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Forma Purified
Isotype IgG