missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Meloxicam-D3 0.1 mg/ml in Acetonitrile, LoGiCal
Specifications
Specifications
| Materiale o nome chimico | Meloxicam D3 (2-methyl D3) |
| CAS | 942047-63-4 |
| Solvente o matrice | Acetonitrile |
| Formula molecolare | C14 2H3 H10 N3 O4 S2 |
| Quantità | 1 mL |
| Sinonimo | Meloxicam D3 (2-methyl D3) |
| InChI Formula | InChI=1S/C14H13N3O4S2/c1-8-7-15-14(22-8)16-13(19)11-12(18)9-5-3-4-6-10(9)23(20,21)17(11)2/h3-7,18H,1-2H3,(H,15,16,19)/i2D3 |
| SMILES | [2H]C([2H])([2H])N1C(=C(O)c2ccccc2S1(=O)=O)C(=O)Nc3ncc(C)s3 |
| IUPAC Name | 4-hydroxy-N-(5-methyl-1,3-thiazol-2-yl)-1,1-dioxo-2-(trideuteriomethyl)-1λ^{6},2-benzothiazine-3-carboxamide |
| Molecular Weight (g/mol) | 354.42 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?