Learn More
Melphalan Hydrochloride, TRC
Specifica
Specifica
| Materiale o nome chimico | Melphalan hydrochloride |
| CAS | 3223-07-2 |
| Formula molecolare | C13 H18 Cl2 N2 O2 . Cl H |
| Quantità | 50 mg |
| Sinonimo | 4-[bis(2-Chloroethyl)amino]-L-phenylalanine Hydrochloride (1:1), Alanine, 3-[p-[bis(2-chloroethyl)amino]phenyl]-, monohydrochloride, L- (8CI), L-Phenylalanine, 4-[bis(2-chloroethyl)amino]-, monohydrochloride (9CI), 4-Bis(2-chloroethyl)amino-L-phenylalanine hydrochloride, Melphalan hydrochloride |
| InChI Formula | InChI=1S/C13H18Cl2N2O2.ClH/c14-5-7-17(8-6-15)11-3-1-10(2-4-11)9-12(16)13(18)19;/h1-4,12H,5-9,16H2,(H,18,19);1H/t12-;/m0./s1 |
| SMILES | Cl.N[C@@H](Cc1ccc(cc1)N(CCCl)CCCl)C(=O)O |
| IUPAC Name | (2S)-2-amino-3-[4-[bis(2-chloroethyl)amino]phenyl]propanoic acid;hydrochloride |
| Molecular Weight (g/mol) | 341.66 |
| Peso formulazione | 340.0512 |