Learn More
Cytiva Membranes
Product Code. 11989217
Change view
Click to view available options
Pore Size:
0.6 μm
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Pore Size Unit Size
11989217 0.6 μm Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11989217 Supplier Cytiva Supplier No. 10409870
  • ONLINE EXCLUSIVE PRICE
    228.00€
    207.00€ / pezzi 100
    Risparmia 21.00€
    9% Sconto
 Restricted Item
Spedizione stimata: 01-12-2026
Specifications
Pore Size 0.6 μm
Sterility Sterile
For Use With (Application) These membranes are used to quantitatively evaluate microorganism counts. The liquid is filtered through a membrane and microorganisms collect on the membrane surface.
Quantity 100Each
Type Membrane
compliance-icons
Product Identifier
  • ME26/31 ST, 0.6µm 47mm
Signal Word
  • Warning
Hazard Category
  • Flammable solid Category 1
  • Flammable solid Category 2
Hazard Statement
  • H228-Flammable solid.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
  • MIXTURE LIST-Contain: nitrocellulose