Learn More
Uvex™ Men's Jacket 7475-Night Blue
Catalog Number 17225921
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
6X-Large
Well thought-out functionality, sporty design, exciting color contrasts, unusual material combinations and a cut that fits like a glove.
  • Jacket uvex suXXeed multifunction
  • Material: 49% PPAN-FR, 42% cotton, 5% Aramid, 3% Polyamid, 1% antistat. fibres, 300 gsm
  • Multifunctional, for hightest protection, soft and skin friendly, integrated stretch inserts, high rise Sleeve construction, reflective inserts
  • Standards: - EN ISO 11611 - EN ISO 11612 - EN 1149-5 - EN 13034 Type 6 - IEC 61482-1-2
  • Sizes: S - 6XL
  • Industrial wash, 60 degree, no bleaching, 2 points tumbler, 2 points ironing
Size 2X-Large
Sesso Men's
Colore Blue
Tipo di chiusura Zipper
Monouso No
Spessore 300 g/m²