Learn More
Meprylcaine Hydrochloride, TRC
Specifica
Specifica
| Materiale o nome chimico | Meprylcaine HCl |
| CAS | 956-03-6 |
| Formula molecolare | C14 H21 N O2 . Cl H |
| Quantità | 100 mg |
| Sinonimo | 2-Methyl-2-(propylamino)-1-propanol Benzoate (Ester) Hydrochloride, 2-Methyl-2-(propylamino)-1-propanol Benzoate Hydrochloride, 2-Methyl-2-propylaminopropyl Benzoate Hydrochloride, Meprylcaine Hydrochloride, Oracaine Hydrochloride |
| InChI Formula | InChI=1S/C14H21NO2.ClH/c1-4-10-15-14(2,3)11-17-13(16)12-8-6-5-7-9-12;/h5-9,15H,4,10-11H2,1-3H3;1H |
| SMILES | Cl.CCCNC(C)(C)COC(=O)c1ccccc1 |
| IUPAC Name | [2-methyl-2-(propylamino)propyl] benzoate;hydrochloride |
| Molecular Weight (g/mol) | 271.78 |
| Peso formulazione | 271.1339 |