Learn More
Trajan Scientific and Medical™ MEPS BINs for XCHANGE/eVol Syringes
Miniaturization of conventional sample preparation devices. Trajan Scientific and Medical MEPS BINs for XCHANGE/eVol Syringes are micro extraction by packed sorbent.
Marca:  Trajan Scientific and Medical™ 2900716
Dettagli aggiuntivi : Peso : 0.01000kg
 vedi altre versioni di questo prodotto
Codice prodotto. 15259253
  • 479.00€ / pezzi 5
Spedizione stimata
25-08-2020
Aggiungi al carrello
Descrizione
Descrizione
  • Save hours in sample preparation
  • Reduce solvent consumption and sample volume
  • Available in reversed phase, normal phase, mixed mode and ion exchange chemistries
Specifica
Specifica
SDVB
Cone Tipped Needle
0.63 mm
MEPS Barrel Insert and Needle (BIN)
XCHANGE/eVol Syringes