Learn More
Trajan Scientific and Medical™ MEPS BINs for XCHANGE/eVol Syringes
Miniaturization of conventional sample preparation devices. Trajan Scientific and Medical MEPS BINs for XCHANGE/eVol Syringes are micro extraction by packed sorbent.
Marca:  Trajan Scientific and Medical™ 2900701
Dettagli aggiuntivi : Peso : 0.01000kg
 vedi altre versioni di questo prodotto
Codice prodotto. 15259243
  • 479.00€ / pezzi 5
Spedizione stimata
24-08-2020
Aggiungi al carrello
Descrizione
Descrizione
  • Save hours in sample preparation
  • Reduce solvent consumption and sample volume
  • Available in reversed phase, normal phase, mixed mode and ion exchange chemistries
Specifica
Specifica
C18
MEPS Barrel Insert and Needle (BIN)
LC Tipped Needle
XCHANGE/eVol Syringes