missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3,6-Trichlorophenol in Iso-propanol 1000μg/mL, Fisher Chemical™
Sfoglia Tutto Fisher Chemical ProdottiSpecifications
Specifications
| Materiale o nome chimico | 2,3,6-Trichloropheno |
| Imballaggio | Ampoule |
| CAS | 933-75-5 |
| Forma fisica | Liquid |
| Formula molecolare | C6H3Cl3O |
| Concentrazione | 1000 μg/mL |
| InChI Key | XGCHAIDDPMFRLJ-UHFFFAOYSA-N |
| Tipo | Organic Single-component Standard Solution |
| SMILES | C1=CC(=C(C(=C1Cl)O)Cl)Cl |
| IUPAC Name | 2,3,6-trichlorophenol |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?