missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Quintozene in Methyl-tert.butylether 1000μg/mL, Fisher Chemical™
Click to view available options
Quantità:
1 mL
Imballaggio:
Ampoule
Specifica
Materiale o nome chimico | Quintozene (Pentachloronitrobenzene) |
Concentrazione | 1000μg/mL |
CAS | 82-68-8 |
Imballaggio | Ampoule |
Quantità | 1 mL |
Tipo | Organic Single-component Standard Solution |
Formula molecolare | C6Cl5NO2 |
InChI Key | LKPLKUMXSAEKID-UHFFFAOYSA-N |
SMILES | C1(=C(C(=C(C(=C1Cl)Cl)Cl)Cl)Cl)[N+](=O)[O-] |
IUPAC Name | 1,2,3,4,5-pentachloro-6-nitrobenzene |
Vedi altri risultati |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'