Learn More
Methoxyflurane, TRC
Specifications
Specifications
| Chemical Name or Material | Methoxyflurane |
| CAS | 76-38-0 |
| Purity Grade Notes | GC |
| Molecular Formula | C3 H4 Cl2 F2 O |
| Quantity | 10 g |
| Synonym | Dichloro-2,2-difluoro-2-methoxyethane, 2,2-Dichloro-1,1-difluoro-1-methoxyethane, 2,2-Dichloro-1,1-difluoroethyl Methyl Ether, Analgizer, Anecotan, DA 759, Inhalan, Methoflurane, Methoxane, Methoxyfluran, Methoxyflurane, Methyl 1,1-difluoro-2,2-dichloroethyl ether, Metofane, Metoxfluran, Metoxifluran, NSC 110432, Penthrane, Pentran, Pentrane |
| InChI Formula | InChI=1S/C3H4Cl2F2O/c1-8-3(6,7)2(4)5/h2H,1H3 |
| SMILES | COC(F)(F)C(Cl)Cl |
| IUPAC Name | 2,2-dichloro-1,1-difluoro-1-methoxyethane |
| Molecular Weight (g/mol) | 164.97 |
| Show More |