Learn More
Methyl 2,3,4-Tri-O-isobutyryl-1-O-trichloroacetimidoyl-Alpha-D-glucopyranuronate, TRC
Codice prodotto. 30480472
Cambia vista
Click to view available options
Quantità:
50 mg
Dimensione della confezione:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
30480472 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 30480472 Fornitore Toronto Research Chemicals N. del fornitore TRCM33093050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifica
Materiale o nome chimico Methyl 2,3,4-Tri-O-isobutyryl-1-O-trichloroacetimidoyl-a-D-g
CAS 150607-96-8
Formula molecolare C21 H30 Cl3 N O10
Quantità 50 mg
InChI Formula InChI=1S/C21H30Cl3NO10/c1-8(2)15(26)31-11-12(32-16(27)9(3)4)14(33-17(28)10(5)6)19(34-13(11)18(29)30-7)35-20(25)21(22,23)24/h8-14,19,25H,1-7H3/t11-,12-,13-,14+,19+/m0/s1
SMILES COC(=O)[C@H]1O[C@H](OC(=N)C(Cl)(Cl)Cl)[C@H](OC(=O)C(C)C)[C@@H](OC(=O)C(C)C)[C@@H]1OC(=O)C(C)C
IUPAC Name methyl (2S,3S,4S,5R,6R)-3,4,5-tris(2-methylpropanoyloxy)-6-(2,2,2-trichloroethanimidoyl)oxyoxane-2-carboxylate
Molecular Weight (g/mol) 562.82
Peso formulazione 561.0935