Learn More
Alfa Aesar™ Methyl 3-aminopyridine-4-carboxylate, 97%
Marca:  Alfa Aesar™ H34007.09
Dettagli aggiuntivi : CAS : 55279-30-6 Peso : 0.01000kg
Quantità 10g
Codice prodotto. 15446907
  • 355.00€ / 10g
Spedizione stimata: 24-06-2021
Identificatori chimici
55279-30-6
152.153
XLQIGLBALJNHKR-UHFFFAOYSA-N
1488824
COC(=O)C1=C(C=NC=C1)N
C7H8N2O2
MFCD02082535
methyl 3-aminoisonicotinate, 3-amino-isonicotinic acid methyl ester, 3-aminopyridine-4-carboxylic acid methyl ester, methyl3-aminoisonicotinate, 3-aminoisonicotinic acid methyl ester, 4-pyridinecarboxylic acid, 3-amino-, methyl ester, 3-amino-4-pyridinecarboxylic acid methyl ester, pubchem9760, acmc-1avm8, methyl 3-amino isonicotinate
methyl 3-aminopyridine-4-carboxylate
Specifica
Methyl 3-aminopyridine-4-carboxylate
10g
C7H8N2O2
methyl 3-aminoisonicotinate, 3-amino-isonicotinic acid methyl ester, 3-aminopyridine-4-carboxylic acid methyl ester, methyl3-aminoisonicotinate, 3-aminoisonicotinic acid methyl ester, 4-pyridinecarboxylic acid, 3-amino-, methyl ester, 3-amino-4-pyridinecarboxylic acid methyl ester, pubchem9760, acmc-1avm8, methyl 3-amino isonicotinate
COC(=O)C1=C(C=NC=C1)N
152.153
152.2
84°C to 86°C
55279-30-6
MFCD02082535
XLQIGLBALJNHKR-UHFFFAOYSA-N
methyl 3-aminopyridine-4-carboxylate
1488824
97%
Videos
SDS Documenti
Certificati