Learn More
Methyl [[4-[2-[[(3-Ethyl-4-methyl-2-oxo-2,3-dihydro-1H-pyrrol-1-yl)carbonyl]amino]ethyl]phenyl]sulphonyl]carbamate (Glimepiride Carbamate), Mikromol™
Descrizione
Methyl [[4-[2-[[(3-Ethyl-4-methyl-2-oxo-2,3-dihydro-1H-pyrrol-1-yl)carbonyl]amino]ethyl]phenyl]sulphonyl]carbamate (Glimepiride Carbamate)
Specifica
Specifica
| Materiale o nome chimico | Glimepiride Carbamate |
| CAS | 119018-30-3 |
| Nomi componenti o analita | Glimepiride Carbamate |
| Shipping Condition | Room Temperature |
| Formula molecolare | C18 H23 N3 O6 S |
| Certificazione/Conformità | ISO 9001 |
| Sinonimo | Glimepiride Imp. C (EP), Glimepiride USP Related Compound C, Glimepiride USP RC C, Methyl [[4-[2-[[(3-Ethyl-4-methyl-2-oxo-2,3-dihydro-1H-pyrrol-1-yl)carbonyl]amino]ethyl]phenyl]sulphonyl]carbamate, Glimepiride Carbamate, Glimepiride Related Compound C, Glimepiride Impurity C |
| InChI Formula | InChI=1S/C18H23N3O6S/c1-4-15-12(2)11-21(16(15)22)17(23)19-10-9-13-5-7-14(8-6-13)28(25,26)20-18(24)27-3/h5-8H,4,9-11H2,1-3H3,(H,19,23)(H,20,24) |
| SMILES | CCC1=C(C)CN(C(=O)NCCc2ccc(cc2)S(=O)(=O)NC(=O)OC)C1=O |
| IUPAC Name | methyl N-[4-[2-[(4-ethyl-3-methyl-5-oxo-2H-pyrrole-1-carbonyl)amino]ethyl]phenyl]sulfonylcarbamate |
| Vedi altri risultati |