Learn More
Methyl 4-(Acetylamino)-2-methoxybenzoate, Mikromol™
Product Code. 17983066
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17983066 100 mg 100mg
30049942 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17983066 Supplier Mikromol Supplier No. MM0073.05
 Restricted Item
Spedizione stimata: 30-11-2026
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Methyl 4-(Acetylamino)-2-methoxybenzoate
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Methyl 4-(Acetylamino)-2-methoxybenzoate
CAS 4093-29-2
Analyte or Component Names Methyl 4-(Acetylamino)-2-methoxybenzoate
Shipping Condition Room Temperature
Molecular Formula C11 H13 N O4
Certifications/Compliance ISO 17025
Synonym Metoclopramide Imp. D (EP), Metoclopramide USP Related Compound D, Metoclopramide USP RC D, Methyl 4-(Acetylamino)-2-methoxybenzoate, Metoclopramide Related Compound D, Metoclopramide Hydrochloride Monohydrate Impurity D, Metoclopramide Impurity D
InChI Formula InChI=1S/C11H13NO4/c1-7(13)12-8-4-5-9(11(14)16-3)10(6-8)15-2/h4-6H,1-3H3,(H,12,13)
SMILES COC(=O)c1ccc(NC(=O)C)cc1OC
IUPAC Name methyl 4-acetamido-2-methoxybenzoate
Molecular Weight (g/mol) 223.23
Quantity 100 mg
Formula Weight 223.0845 g/mol