missing translation for 'onlineSavingsMsg'
Learn More

Methyl Cyano(2-nitrophenyl)acetate, TRC

Product Code. 30468740
Change view
Click to view available options
Quantità:
2 g
Unit Size:
2g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantità Unit Size
30468740 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30468740 Supplier Toronto Research Chemicals Supplier No. TRCM2945502G
 Articolo soggetto a restrizioni
Spedizione stimata: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Materiale o nome chimico Methyl Cyano(2-nitrophenyl)acetate
CAS 113772-13-7
Formula molecolare C10 H8 N2 O4
Quantità 2 g
InChI Formula InChI=1S/C10H8N2O4/c1-16-10(13)8(6-11)7-4-2-3-5-9(7)12(14)15/h2-5,8H,1H3
SMILES COC(=O)C(C#N)c1ccccc1[N+](=O)[O-]
IUPAC Name methyl 2-cyano-2-(2-nitrophenyl)acetate
Molecular Weight (g/mol) 220.18
Peso formulazione 220.0484
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.