Learn More
Methyl Isofenphos (Technical Grade), TRC
Specifica
Specifica
| Materiale o nome chimico | Isofenphos-methyl |
| CAS | 99675-03-3 |
| Formula molecolare | C14 H22 N O4 P S |
| Quantità | 250 mg |
| Sinonimo | Isofenphos-methyl, Isophenphos-methyl, ISP-methyl, Jiajiyiliulin, Methyl isofenphos, Methyl isophenphos, Methyl-ISP, Yulioulin-methyl |
| InChI Formula | InChI=1S/C14H22NO4PS/c1-10(2)15-20(21,17-5)19-13-9-7-6-8-12(13)14(16)18-11(3)4/h6-11H,1-5H3,(H,15,21) |
| SMILES | COP(=S)(NC(C)C)Oc1ccccc1C(=O)OC(C)C |
| IUPAC Name | propan-2-yl 2-[methoxy-(propan-2-ylamino)phosphinothioyl]oxybenzoate |
| Molecular Weight (g/mol) | 331.37 |
| Peso formulazione | 331.1007 |