Learn More
Methyl Orange Mixed Indicator Solution, Honeywell Fluka™
Acid-base indicator
Marca: Honeywell Fluka 32940-100ML
Dettagli aggiuntivi : Peso : 0.10000kg
Descrizione
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifica
Methyl Orange Mixed Indicator Solution | |
NONH for all modes of transport | |
Glass bottle | |
100 mL |
MFCD00007502 | |
3659741 | |
≥0.999g/mL at 20°C |