Learn More
Methyl Salicylate Beta-D-O-Glucuronide Triacetate Methyl Ester, TRC
Specifica
Specifica
| Materiale o nome chimico | Methyl Salicylate beta-D-O-Glucuronide Triacetate Methyl Ester |
| CAS | 101231-54-3 |
| Formula molecolare | C21H24O12 |
| Quantità | 10 mg |
| Sinonimo | 2-(Methoxycarbonyl)phenyl beta-D-Glucopyranosiduronic Acid Methyl Triacetate Ester, O-carboxyphenyl beta-D-Glucopyranosiduronic alphacid Triacetate Dimethyl Ester. |
| InChI Formula | InChI=1S/C21H24O12/c1-10(22)29-15-16(30-11(2)23)18(31-12(3)24)21(33-17(15)20(26)28-5)32-14-9-7-6-8-13(14)19(25)27-4/h6-9,15-18,21H,1-5H3/t15-,16-,17-,18+,21+/m0/s1 |
| SMILES | COC(=O)[C@H]1O[C@@H](Oc2ccccc2C(=O)OC)[C@H](OC(=O)C)[C@@H](OC(=O)C)[C@@H]1OC(=O)C |
| IUPAC Name | methyl (2S,3S,4S,5R,6S)-3,4,5-triacetyloxy-6-(2-methoxycarbonylphenoxy)oxane-2-carboxylate |
| Molecular Weight (g/mol) | 468.41 |
| Peso formulazione | 468.1268 |