missing translation for 'onlineSavingsMsg'
Learn More

Methyle4-methoxythiophene-3-carboxylate, 97%, Thermo Scientific™

Catalog Number p-2705383
Click to view available options
Quantità:
250 mg
1 g
5 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
65369-22-4
C7H8O3S
172.198
MFCD00067995
LQIVVUILGDRRHE-UHFFFAOYSA-N
methyl 4-methoxy-3-thiophenecarboxylate, 4-methoxy-thiophene-3-carboxylic acid methyl ester, 3-thiophenecarboxylic acid, 4-methoxy-, methyl ester, maybridge1_004625, ksc495s3h, methyl 4-methoxythiophene-3-arboxylate, 3-methoxythiophene-4-carboxylic acid methyl ester, 3-thiophenecarboxylicacid,4-methoxy-,methyl ester, 4-methoxy-3-thiophenecarboxylic acid methyl ester, 3-thiophenecarboxylicacid, 4-methoxy-, methyl ester
2778377
methyl 4-methoxythiophene-3-carboxylate
COC1=CSC=C1C(=O)OC
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
65369-22-4
C7H8O3S
172.198
MFCD00067995
LQIVVUILGDRRHE-UHFFFAOYSA-N
methyl 4-methoxy-3-thiophenecarboxylate, 4-methoxy-thiophene-3-carboxylic acid methyl ester, 3-thiophenecarboxylic acid, 4-methoxy-, methyl ester, maybridge1_004625, ksc495s3h, methyl 4-methoxythiophene-3-arboxylate, 3-methoxythiophene-4-carboxylic acid methyl ester, 3-thiophenecarboxylicacid,4-methoxy-,methyl ester, 4-methoxy-3-thiophenecarboxylic acid methyl ester, 3-thiophenecarboxylicacid, 4-methoxy-, methyl ester
2778377
methyl 4-methoxythiophene-3-carboxylate
COC1=CSC=C1C(=O)OC
Materiale o nome chimico Methyl 4-methoxythiophene-3-carboxylate
CAS min. % 97.0
Intervallo percentuale saggio 97%
Imballaggio Amber glass bottle
Quantità 250 mg
Peso formulazione 172.2
Percent Purity 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'