missing translation for 'onlineSavingsMsg'
Learn More

Methyltriphenoxyphosphonium iodide, 95%, Thermo Scientific Chemicals

Catalog Number p-3779250
25.13 EUR valido fino al 2024-03-29
Usa il codice promo "21615" per avere il tuo prezzo promozionale.


Click to view available options
Quantità:
5 g
50 g
Imballaggio:
Glass bottle
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
17579-99-6
C19H18IO3P
452.22
MFCD00011911
VKTOBGBZBCELGC-UHFFFAOYSA-M
methyltriphenoxyphosphonium iodide, methyltriphenoxyphosphanium iodide, acmc-20ake2, triphenyl phosphite methoiodide, methyl triphenoxyphosphonium iodide, methyl triphenoxy phosphanium iodide, methyltriphenoxyphosphorus 1+ iodide, phosphorus 1+ ,methyltriphenoxy-, iodide 1:1 , t-4
2735090
methyl(triphenoxy)phosphanium;iodide
C[P+](OC1=CC=CC=C1)(OC2=CC=CC=C2)OC3=CC=CC=C3.[I-]
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
17579-99-6
C19H18IO3P
452.22
MFCD00011911
VKTOBGBZBCELGC-UHFFFAOYSA-M
methyltriphenoxyphosphonium iodide, methyltriphenoxyphosphanium iodide, acmc-20ake2, triphenyl phosphite methoiodide, methyl triphenoxyphosphonium iodide, methyl triphenoxy phosphanium iodide, methyltriphenoxyphosphorus 1+ iodide, phosphorus 1+ ,methyltriphenoxy-, iodide 1:1 , t-4
2735090
methyl(triphenoxy)phosphanium;iodide
C[P+](OC1=CC=CC=C1)(OC2=CC=CC=C2)OC3=CC=CC=C3.[I-]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico Methyltriphenoxyphosphonium iodide
CAS min. % 94.0
CAS max % 100.0
Punto di fusione 141.0°C to 146.0°C
Intervallo percentuale saggio 94% min. (Argentometry)
Imballaggio Glass bottle
Quantità 5 g
Beilstein 06, 177
Fieser 01,1249; 02,446; 04,557; 06,649; 08,354
Informazioni di solubilità Solubility in water: negligible. Other solubilities: soluble in acetone,diethyl ether
Peso formulazione 452.22
Percent Purity 95%
Vedi altri risultati Mostra meno risultati
compliance-icons
Identificativo prodotto
  • Methyltriphenoxyphosphonium iodide
Avvertenza
  • Attenzione
Categoria di pericolo
  • Lesione oculare grave/irritazione oculare Categoria 2
  • Corrosione/irritazione della pelle Categoria 2
  • Tossicità specifica per organi bersaglio (esposizione singola) Categoria 3
Indicazioni di pericolo
  • H319-Provoca grave irritazione oculare.
  • H335-Può irritare le vie respiratorie.
  • H315-Provoca irritazione cutanea.
Consigli di prudenza
  • P261-Evitare di respirare la polvere/i fumi/i gas/la nebbia/i vapori/gli aerosol.
  • P280-Indossare guanti/indumenti protettivi/Proteggere gli occhi/il viso.
  • P302+P352-IN CASO DI CONTATTO CON LA PELLE: lavare abbondantemente con acqua/.
  • P305+P351+P338-IN CASO DI CONTATTO CON GLI OCCHI: sciacquare accuratamente per parecchi minuti. Togliere le eventuali lenti a contatto se è agevole farlo. Continuare a sciacquare.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'