Learn More
Metro™ High-Density qwikTRAK™ Mobile Unit Kits
Descrizione
- For use with Metro Super Erecta™, Super Erecta Pro™ Super Adjustable™ and MetroMax iQ™ shelving systems
- Kit includes four 74 in. (188cm) high mobile posts, four V-groove casters; one stainless steel center channel caster assembly, donut bumpers, tie bar brackets and other hardware
Order Info
Order one kit per mobile system. Shelves are sold separately.
Specifica
Specifica
| Product Type | Mobile Unit Kit |
| Finitura | Chrome |
| Da utilizzare con (apparecchiatura) | Metro Super Erecta, Super Adjustable Shelving System |
| Include | Four 74 in. high mobile posts, four V-groove casters, one Stainless Steel center channel caster assembly, donut bumpers, tie bar brackets and other hardware |
| Peso (sistema imperiale) | 25 lb. |
| Peso (sistema metrico) | 11.4 kg |
| Materiale | Steel |