Learn More
Mettler Toledo™ pH Plug and Play Glass Titration Electrodes
Product Code. 15137132 Shop All Mettler Toledo Products
Change view
Click to view available options
Diameter (Metric):
12 mm
Filling Solution Type:
1 mol/L LiCl
3 mol/L KCl
KCl gel, 1 mol/L KCl
Junction:
Ceramic Pin
Ground Glass
Ground Glass, Ceramic Pin
Moveable Tefzel
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Filling Solution Type Junction Unit Size
15137132 12 mm KCl gel, 1 mol/L KCl Ground Glass, Ceramic Pin Each
11347260 12 mm 3 mol/L KCl Ground Glass Each
12724922 12 mm 1 mol/L LiCl Moveable Tefzel Each
11337260 12 mm 3 mol/L KCl Ceramic Pin Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15137132 Supplier Mettler Toledo™ Supplier No. 51109506
  • ONLINE EXCLUSIVE PRICE
    1560.70€
    1229.39€ / pezzi 1
    Risparmia 331.31€
    21% Sconto
 Restricted Item
Spedizione stimata: 21-01-2027
Includes
25mL of reference electrolyte, detachable cable, and quality certificate with unique serial number (all models except DGi114SC, which contains sensor chip)
For direct pH measurements and acid/base titrations in aqueous or non-aqueous media. Mettler Toledo pH Plug and Play Glass Titration Electrodes includes a Plug and Play sensor with integrated sensor chip for secure handling.
  • ARGENTHAL™ reference system present
  • Features refillable reference electrolyte
  • Shaft material is made of glass
  • Length, 150mm; Diameter, 12mm
Model DGi101-SC and DGi102-Mini
  • U-glass type membrane glass
  • Diaphragm is ceramic pin
  • Silver ion trap to ensure that the electrolyte is absolutely free from silver ions
DGi111-SC
  • HA-taped type membrane glass
  • Diaphragm is ceramic pin
  • Silver ion trap to ensure that the electrolyte is absolutely free from silver ions
DGi113-SC
  • A41 cylindrical type membrane glass
  • Movable PTFE sleeve junction
  • Integrated glass sleeve for secure fixation in the titration head
DGi115-SC
  • HA-cylindrical type membrane glass
  • Fixed ground glass sleeve junction
  • Silver ion trap to ensure that the electrolyte is absolutely free from silver ions
DGi114-SC
  • HA-cylindrical type membrane glass
  • Movable PTFE sleeve junction
DGi116-Solvent
  • Spherical A41 glass membrane of low resistance for fast and sensitive pH response and chemical resistance
  • Easy-to-clean glass sleeve junction providing a uniform electrolyte flow
  • Gold shielding for efficient electric and static shielding
  • Movable ground glass sleeve junction
DGi117-Water
  • Double junction construction
  • Fixed ground-glass sleeve junction
  • Features Pt1000 temperature sensor
  • Integrated glass sleeve for secure fixation in the titration head
  • Gold shielding against electrostatic effects
  • LoT spherical type membrane glass
  • Spherical glass membrane of low resistance
  • MultiPin™ screw cap for secure connection of combined electrode and temperature probe cable
TRUSTED_SUSTAINABILITY
Specifications
Connector Types S12
Junction Ground Glass, Ceramic Pin
Material Glass
pH Range 1 to 11
Temperature (Metric) Operating 0°C to 100°C
Type pH Plug and Play Electrode
Filling Solution Type KCl gel, 1 mol/L KCl
For Use With (Application) For direct pH measurements and acid/base titrations
Includes Integrated temperature sensor, for aqueous media
Internal Reference ARGENTHAL
Membrane LoT Glass
Model DGi117-Water
Precision High
Temperature (English) Operating 32°F to 212°F
Temperature (Metric) Range 0°C to 100°C
Length (English) 7.28 in.
Length (Metric) 185.1 mm
Diameter (English) 0.47 in.
Diameter (Metric) 12 mm