Learn More
Mettler Toledo™ Replacement Parts for V20 Karl Fischer™ Autotitrator
Shop All Mettler Toledo ProductsDescription
- Accessory for DV1000 titration burettes
- For titrant protection from moisture or CO2
- With cover
Specifications
Specifications
| Type | Drying Cup |
| For Use With (Application) | Water content determination |
| Weight (Metric) | 453.5 g |
Product Title
Haben Sie Verbesserungsvorschläge?