Learn More
Mettler Toledo™ Tweezers for Analytical and Calibration Weights
Codice prodotto. 10324225 Sfoglia Tutto Mettler Toledo Prodotti
Cambia vista
Click to view available options
Da utilizzare con (applicazione):
1mg to 50g Weights
Dimensione della confezione:
Pezzo
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Da utilizzare con (applicazione) unitSize
10324225 1mg to 50g Weights Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 10324225 Fornitore Mettler Toledo™ N. del fornitore 15900
Accessories for Weights. Ergonomic Weight Handling - Ergonomic tweezers and weight forks are made of clean room suitable materials and allow professional weight handling and balance testing.
Constructed of metal with carbon reinforced nylon.
TRUSTED_SUSTAINABILITY
Specifica
Product Type Tweezers
Materiale Metal with Carbon Reinforced Nylon
Da utilizzare con (applicazione) 1mg to 50g Weights
Quantità Each