missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MGAT5B Polyclonal Antibody

Descrizione
Immunogen sequence: RTKRLTAQWAL AAQRLAQKLG ATQRDQKQIL VHIGFLTEES GDVFSPRVLK GGPLGEMVQW ADILTALYVL GHGLRVTVSL KELQSNLGVP PGRGSCPLTM PLPFDLIYTD YHGLQQMKRH MGLSFKKYRC RIRVID Highest antigen sequence identity to the following orthologs - mouse 89%, rat 88%.
Glycosyltransferase that acts on alpha-linked mannose of N-glycans and O-mannosyl glycans. Catalyzes the transfer of N-acetylglucosamine (GlcNAc) to the beta 1-6 linkage of the mannose residue of GlcNAcbeta1,2-Manalpha on both the alpha1,3- and alpha1,6-linked mannose arms in the core structure of N-glycan. Also acts on the GlcNAcbeta1,2-Manalpha1-Ser/Thr moiety, forming a 2,6-branched structure in brain O-mannosyl glycan. Plays an active role in modulating integrin and laminin-dependent adhesion and migration of neuronal cells via its activity in the O-mannosyl glycan pathway.
Specifica
Specifica
| Antigene | MGAT5B |
| Applicazioni | Immunocytochemistry |
| Classificazione | Polyclonal |
| Concentrazione | 0.05 mg/mL |
| Coniugato | Unconjugated |
| Formulazione | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | MGAT5B |
| N. accesso geni | Q3V5L5 |
| Alias gene | Alpha-1,6-mannosylglycoprotein 6-beta-N-acetylglucosaminyltransferase B; alpha-mannoside beta-1,6-N-acetylglucosaminyltransferase B; beta(1,6)-N-acetylglucosaminyltransferase V; beta1,6-N-acetylglucosaminyltransferase IX (GnT-IX); C330018B01; GlcNAc-T Vb; GnT-IX; GNT-Vb; hGnTVb; KIAA2008; mannoside acetylglucosaminyltransferase 5, isoenzyme B; mannoside acetylglucosaminyltransferase 5B; mannosyl (alpha-1,6-)-glycoprotein beta-1,6-N-acetyl-glucosaminyltransferase, isoenzyme B; mannosyl (alpha-1,6-)-glycoprotein beta-1,6-N-acetyl-glucosaminyltransferase, isozyme B; Mgat5b; mGnTVB; N-acetylglucosaminyltranferase VB; N-acetylglucosaminyltransferase IX; N-acetylglucosaminyl-transferase Vb; UDP-N-acetylglucosamine: alpha1,6-D-mannoside beta1,6-N-acetylglucosaminyltransferase b |
| Simboli geni | MGAT5B |
| Vedi altri risultati |
Titolo del prodotto
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.
Individuate un'opportunità di miglioramento?