Learn More
Hecht Karl™ Micro Hematocrit Capillaries
Product Code. 10718472
Change view
Click to view available options
Quantity:
10Pack
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10718472 10Pack Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10718472 Supplier Hecht Karl™ Supplier No. 40563010
  • ONLINE EXCLUSIVE PRICE
    4.47€
    2.35€ / pezzi 100
    Risparmia 2.12€
    47% Sconto
View all promotions
Spedizione stimata: 16-12-2026
Order Info
LengthMetricTube
Specifications
Product Type Capillary Tube
Diameter (Metric) Inner 1.1 to 1.2 mm
For Use With (Equipment) Hematocrit Centrifuges
Quantity 10Pack
For Use With (Application) Hematocrit centrifuges
Length (Metric) 75 mm