Learn More
Emma Safety Footwear Micro Safety Shoes
Unique microfiber upper is fully washable. SmartecAir™ provides fresh dry feet. Protective toe cap prevents staining and is lubricant and fat resistant. The sole was developed for use on hard tiled floors and other easy-to-clean floors used in high-tech and cleanroom environments.
Marca:  Emma Safety Footwear 500540-38
Dettagli aggiuntivi : Peso : 1.25400kg
Codice prodotto. 15938451
Quantità Prezzo
1 79.25€ / 1 paio
10 74.61€ / 1 paio
Spedizione stimata
25-01-2021
Aggiungi al carrello
Descrizione
Descrizione
  • S2-Safety shoe for wet working conditions
  • Microfiber construction
  • GripForce™ Easy Twist Outsole
  • ESD certified
  • Chrome-free lining
  • Hydro-Tec™ Sanitized Silver for drier, fresher feet
  • Sanitized
Specifica
Specifica
EN ISO 20345:2011, EN 12568:2010
White
Hydro-Tec™ Sanitized Silver
38
Elastic
Micro
Microfiber
Steel