Learn More
Microbiologics™ Epower™ Pseudomonas aeruginosa, ATCC™ 9027™†
Description
Includes
Product warranty and peel-off certificate of assay
- Biosafety level: 2
- Mean Assay Value: 1.0-9.9E+07 CFU per pellet
Order Info
Available in vials containing 10 lyophilized pellets, with a range of concentration from 102 to 109 per pellet
Spécification
Spécification
| ATCC® | 9027 |
| Livello di biosicurezza | 2 |
| Forma | Lyophilized Pellet |
| Product Type | QC Microorganism |
| Da utilizzare con (applicazione) | Lyophilized pellets add convenience to special projects and internal proficiency plans. |
| Include | Product warranty and peel-off certificate of assay |
| Valore medio saggio | 0-9.9E+07 CFU per pellet |
| Tipo di imballaggio | Vial |
| Quantità | 10/Pk. |
| Specie | Pseudomonas aeruginosa |
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. Microbiologics Inc. is licensed to use these trademarks and to sell products derived from ATCC™ cultures.
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.