Learn More
Microbiologics™ Epower™ Staphylococcus aureus subsp. Aureus, ATCC™ 6538™†
Product Code. 11979577
Encompass_Preferred
Change view
Click to view available options
Mean Assay Value:
1.0-9.9E+07 CFU per pellet
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mean Assay Value Unit Size
11979577 1.0-9.9E+07 CFU per pellet Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11979577 Supplier Microbiologics™ Supplier No. 0485E7
 Restricted Item
Spedizione stimata: 14-12-2026
Includes
Product warranty and peel-off certificate of assay
Lyophilized pellets add convenience to special projects and internal proficiency plans
  • Biosafety Level: 2
  • Mean Assay Value: 1.0-9.9E+07 CFU per pellet
Order Info
Available in vials containing 10 lyophilized pellets, with a range of concentration from 102 to 109 per pellet
Specifications
ATCC® 6538
Biosafety Level 2
Form Lyophilized Pellet
Product Type Epower
For Use With (Application) Lyophilized pellets add convenience to special projects and internal proficiency plans.
Includes Product warranty and peel-off certificate of assay
Mean Assay Value 1.0-9.9E+07 CFU per pellet
Packaging Type Vial
Quantity 10/Pk.
Species Staphylococcus aureus
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. Microbiologics Inc. is licensed to use these trademarks and to sell products derived from ATCC™ cultures.